missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APBB3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552167
This item is not returnable.
View return policy
Description
Immunogen sequence: AIILVNCDDD LWGDHSLEVE AGLPPGWRKI HDAAGTYYWH VPSGSTQWQR PTWELGDAED PGTGTEGIWG LRPPKGRSFS SLESSLDRSN SLSWYGGESY IQSMEPGAKC FAVRSL Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.
The protein encoded by this gene is a member of the APBB protein family. It is found in the cytoplasm and binds to the intracellular domain of the Alzheimer's disease beta-amyloid precursor protein (APP) as well as to other APP-like proteins. It is thought that the protein encoded by this gene may modulate the internalization of APP. Multiple transcript variants encoding several different isoforms have been found for this gene.
Specifications
| APBB3 | |
| Polyclonal | |
| Unconjugated | |
| APBB3 | |
| amyloid beta (A4) precursor protein-binding, family B, member 3; amyloid beta A4 precursor protein-binding family B member 3; amyloid beta precursor protein binding family B member 3; amyloid precursor interacting protein; Amyloid-beta A4 precursor protein-binding family B member 3; APBB3; Fe65L2; fe65-like protein 2; Protein Fe65-like 2; Rirl2; SRA; TR2S | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10307 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95704 | |
| APBB3 | |
| Recombinant protein corresponding to Human APBB3. Recombinant protein control fragment (Product #RP-88822). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction