missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APC Antibody (OTI2B8), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70183AF350
This item is not returnable.
View return policy
Description
APC Monoclonal specifically detects APC in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| APC | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| APC | |
| Recombinant protein fragment corresponding to amino acids 955-1382 of human APC (NP_001120983) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2B8 | |
| Western Blot, Immunohistochemistry | |
| adenomatosis polyposis coli, adenomatosis polyposis coli tumor suppressor, adenomatous polyposis coli, adenomatous polyposis coli protein, BTPS2, Deleted in polyposis 2.5, DP2.5DP2, DP3, FPC, GS, Protein APC | |
| Mouse | |
| Affinity Purified | |
| Cancer, Cell Biology, Cell Cycle and Replication, Metastasis, Tumor Suppressors, Wnt Signaling Pathway | |
| 324.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction