missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APC-conjugated IL-6 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
€ 133.74 - € 361.81
Specifications
| Antigen | IL-6 |
|---|---|
| Clone | 4G3E6 |
| Concentration | 200 μg/mL |
| Applications | Not Available |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234080
|
Proteintech
APC-69001-100UL |
100 μL |
€ 361.81
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233980
|
Proteintech
APC-69001-20UL |
20 μL |
€ 133.74
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
APC-conjugated IL-6 Mouse Monoclonal Antibody, Proteintech shows reactivity in Human samples.Specifications
| IL-6 | |
| 200 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 3569 | |
| RecombinantProtein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 4G3E6 | |
| Not Available | |
| APC | |
| Mouse | |
| Human | |
| BSF2; HGF; HSF; IFNB2; IL-6 | |
| IgG1 | |
| Protein G purification | |
| Conjugated Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title