missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APC Monoclonal Antibody (OTI1B12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803682S
This item is not returnable.
View return policy
Description
APC Monoclonal antibody specifically detects APC in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| APC | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O43187 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3656 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| OTI1B12 | |
| Unconjugated | |
| IRAK2 | |
| adenomatosis polyposis coli; adenomatosis polyposis coli tumor suppressor; adenomatous polyposis coli; adenomatous polyposis coli protein; AI047805; APC; APC, WNT signaling pathway regulator; AU020952; AW124434; BTPS2; CC1; Deleted in polyposis 2.5; DP2; DP2.5; DP3; FAP; FPC; GS; mAPC; Min; multiple intestinal neoplasia; PPP1R46; Protein APC; protein phosphatase 1, regulatory subunit 46; RATAPC; truncated adenomatosis polyposis coli; WNT signaling pathway regulator | |
| Human recombinant protein fragment corresponding to amino acids 362-625 of human IRAK2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction