missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APC-PCI Complex Antibody (M36), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-41380H
This item is not returnable.
View return policy
Description
APC-PCI Complex Monoclonal antibody specifically detects APC-PCI Complex in Human samples. It is validated for ELISA
Specifications
| APC-PCI Complex | |
| Monoclonal | |
| HRP | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human | |
| Purified |
| ELISA | |
| M36 | |
| PBS | |
| Mixture of PCI in complex with PSA (approx. 80%) and PCI cleaved from such a complex (approx. 20%). | |
| 100 μg | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction