missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apc1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-77375-0.025mg
This item is not returnable.
View return policy
Description
Apc1 Polyclonal specifically detects Apc1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Apc1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| anaphase promoting complex subunit 1, anaphase-promoting complex 1 (meiotic checkpoint regulator), anaphase-promoting complex subunit 1, APC1Testis-specific gene 24 protein, Cyclosome subunit 1, Mitotic checkpoint regulator, TSG24MCPR | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2μg/mL, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| Q9H1A4 | |
| ANAPC1 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human APC1. The immunogen is located within amino acids 1810 - 1860 of APC1. Amino Acid Squence: SRSHQERLQNHPKRGLF | |
| 0.025 mg | |
| Cell Biology, Cell Cycle and Replication, Ubiquitin Proteasome Pathway | |
| 64682 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction