missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APC1 Monoclonal Antibody (7G9B3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531621
This item is not returnable.
View return policy
Description
MA5-31621 has been tested in indirect ELISA.
Cell cycle regulated protein ubiquitination and degradation within subcellular domains is thought to be essential for the normal progression of mitosis. APC1, also known as mitotic checkpoint regulator (MCPR), is a highly conserved component of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. APC/C is responsible for degrading anaphase inhibitors, mitotic cyclins, and spindle-associated proteins ensuring that events of mitosis take place in proper sequence. The individual APC/C components mRNA and protein levels are expressed at approximately the same levels in most tissues and cell lines, suggesting that they perform their functions as part of a complex.
Specifications
| APC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q9H1A4 | |
| ANAPC1 | |
| Purified recombinant fragment of human ANAPC1 (AA: 12-155) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
| 7G9B3 | |
| Unconjugated | |
| ANAPC1 | |
| 2610021O03Rik; AI047775; AI853536; ANAPC1; anaphase promoting complex subunit 1; anaphase-promoting complex 1 (meiotic checkpoint regulator); anaphase-promoting complex subunit 1; APC1; AW547281; Cyclosome subunit 1; Mcpr; meiotic check point regulator; mitotic checkpoint regulator; Testis-specific gene 24 protein; TSG24 | |
| Mouse | |
| Protein G | |
| RUO | |
| 64682 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction