missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apc10 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77157PCP
This item is not returnable.
View return policy
Description
Apc10 Polyclonal antibody specifically detects Apc10 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Apc10 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| anaphase promoting complex subunit 10, anaphase-promoting complex subunit 10, APC10DOC1, Cyclosome subunit 10, DKFZP564L0562 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the center of human APC10. The immunogen is located within amino acids 60 - 110 of APC10. Amino Acid Squence: IYADYKSDESYTPSK | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Ubiquitin Proteasome Pathway | |
| 10393 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction