missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apc2 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77376B
This item is not returnable.
View return policy
Description
Apc2 Polyclonal antibody specifically detects Apc2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Apc2 | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| adenomatosis polyposis coli 2, adenomatous polyposis coli like, adenomatous polyposis coli protein 2, APCL | |
| Antibody was raised against an 18 amino acid synthetic peptide near the center of human APC2. The immunogen is located within amino acids 240 - 290 of APC2. Amino Acid Squence: TEVPTHPEDGTPQPGNSK | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Ubiquitin Proteasome Pathway | |
| 10297 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunofluorescence, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Biotin | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction