missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apc4 Antibody (CIV1.1), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50105AF594
This item is not returnable.
View return policy
Description
Apc4 Monoclonal antibody specifically detects Apc4 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Apc4 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| anaphase promoting complex subunit 4, anaphase-promoting complex subunit 4, APC4Cyclosome subunit 4 | |
| C-terminus of Apc4 | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 29945 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| CIV1.1 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction