missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apc5 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-55319G
This item is not returnable.
View return policy
Description
Apc5 Polyclonal antibody specifically detects Apc5 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Apc5 | |
| Polyclonal | |
| Western Blot | |
| anaphase promoting complex subunit 5, anaphase-promoting complex subunit 5, APC5Cyclosome subunit 5 | |
| The immunogen recognized by this antibody maps to a region between residue 705 and the C-terminus (residue 755) of human anaphase promoting complex subunit 5 using the numbering given in entry NP_057321.2 (GeneID 51433). | |
| 0.1 mL | |
| Cell Cycle and Replication, DNA Repair | |
| 51433 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction