missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apc7 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77379AF594
This item is not returnable.
View return policy
Description
Apc7 Polyclonal antibody specifically detects Apc7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Apc7 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 594 | |
| anaphase promoting complex subunit 7, anaphase-promoting complex subunit 7, APC7Cyclosome subunit 7 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human APC7. The immunogen is located within amino acids 520 - 570 of APC7. Amino Acid Squence: LDPNDQKSLEGMQKMEME | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 51434 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction