missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APCDD1 Antibody (2584B) [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB10501JF635
This item is not returnable.
View return policy
Description
APCDD1 Monoclonal antibody specifically detects APCDD1 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| APCDD1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| adenomatosis polyposis coli down-regulated 1, Adenomatosis polyposis coli down-regulated 1 protein, B7323, DRAPC1, protein APCDD1 | |
| Mouse myeloma cell line, NS0-derived human APCDD1, Leu27-His492, Accession # Q8J025 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 2584B | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Cancer | |
| 147495 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction