missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ APCDD1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16501432
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16501432 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16501432 Supplier Invitrogen™ Supplier No. PA598605

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

APCDD1 Polyclonal Antibody for ELISA, IHC (P), Western Blot

APCDD1 (Drapc1) is a novel protein that has been shown to be a target of Wnt/beta-catenin signaling pathway in cancer cell lines. APCDD1 is overexpressed in colorectal carcinogenesis and is deregulated in CTNNB1-mutated Wilms tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APCDD1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene APCDD1
Gene Accession No. Q8J025
Gene Alias AB023957; adenomatosis polyposis coli down-regulated 1; adenomatosis polyposis coli down-regulated 1 protein; Adenomatosis polyposis coli down-regulated 1 protein homolog; APC down-regulated 1; apcdd1; APCDD1 (HGNC:15718); APCDD1 homologue; AU041258; B7323; cAPCDD1; Drapc1; EIG180; ethanol induced gene product EIG180; FP7019; HHS; HTS; hypoptrichosis simplex; HYPT1; Pgo1; primglo1; protein APCDD1; Protein primglo1
Gene Symbols APCDD1
Host Species Rabbit
Immunogen Recombinant Human Protein APCDD1 protein (22-288AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147495
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.