missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Apelin receptor Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114830
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Antibody detects endogenous levels of total AGTRL1.
APLNR (Apelin Receptor) is a member of the G protein-coupled receptor gene family. The encoded protein is related to the angiotensin receptor, but is actually an apelin receptor that inhibits adenylate cyclase activity and plays a counter-regulatory role against the pressure action of angiotensin II by exerting hypertensive effect. It functions in the cardiovascular and central nervous systems, in glucose metabolism, in embryonic and tumor angiogenesis and as a human immunodeficiency virus (HIV-1) coreceptor. Two transcript variants resulting from alternative splicing have been identified.
Spezifikation
| Apelin receptor | |
| Polyclonal | |
| Unconjugated | |
| Aplnr | |
| AGTRL1; angiotensin II receptor-like 1; angiotensin receptor-like 1; Apelin receptor; APJ; APJ (apelin) receptor; APJ receptor; APJR; Aplnr; B78; G protein-coupled receptor APJ; GPCR34; G-protein coupled receptor APJ; G-protein coupled receptor HG11; HG11; HG11 orphan receptor; MSR; msr/apj | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 187, 23796, 83518 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P35414, Q9JHG3, Q9WV08 | |
| Aplnr | |
| A synthesized peptide derived from human APLNR(Accession P35414), corresponding to amino acid residues A147-W197. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur