missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APEX1, Mouse anti-Human, Clone: 13B8E5C2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB2372.100ug
This item is not returnable.
View return policy
Specifications
| APEX1 | |
| Monoclonal | |
| Unconjugated | |
| In PBS (0.05% sodium azide) | |
| APE/APE-1/APE1/APEN/APEX/APX/HAP1/REF-1/REF1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody is specific to the human APE/ref-1 protein. | |
| Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 13B8E5C2 | |
| Immunocytochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) The optimal working dilution should be determined by the end user. | |
| APEX nuclease (multifunctional DNA repair enzyme) 1 | |
| APEX1 | |
| Human APEX1. | |
| RUO | |
| 328 | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction