missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ API5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598281
This item is not returnable.
View return policy
Description
API5 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
This gene encodes an apoptosis inhibitory protein whose expression prevents apoptosis after growth factor deprivation. This protein suppresses the transcription factor E2F1-induced apoptosis and also interacts with, and negatively regulates Acinus, a nuclear factor involved in apoptotic DNA fragmentation. Its depletion enhances the cytotoxic action of the chemotherapeutic drugs. Multiple alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| API5 | |
| Polyclonal | |
| Unconjugated | |
| API5 | |
| AAC11; AAC-11; AI196452; antiapoptosis clone 11 protein; API5; API-5; Apoptosis inhibitor 5; cell migration-inducing gene 8 protein; Fibroblast growth factor 2-interacting factor; fibroblast growth factor 2-interacting factor 2; FIF; FIF C1; MIG8; migration-inducing protein MIG8; Protein XAGL | |
| Rabbit | |
| Protein G | |
| RUO | |
| 11800, 8539 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.61 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O35841, Q9BZZ5 | |
| API5 | |
| Recombinant Human Apoptosis inhibitor 5 protein (396-524AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction