missing translation for 'onlineSavingsMsg'
Learn More
Learn More
API5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25689-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes an apoptosis inhibitory protein whose expression prevents apoptosis after growth factor deprivation. This protein suppresses the transcription factor E2F1-induced apoptosis and also interacts with, and negatively regulates Acinus, a nuclear factor involved in apoptotic DNA fragmentation. Its depletion enhances the cytotoxic action of the chemotherapeutic drugs. Multiple alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
| API5 | |
| Polyclonal | |
| Unconjugated | |
| API5 | |
| AAC 11, AAC11, Antiapoptosis clone 11 protein, API 5, API5, apoptosis inhibitor 5, FIF, Protein XAGL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11800, 8539 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O35841, Q9BZZ5 | |
| Api5 | |
| API5 Fusion Protein Ag22524 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction