Learn More
APIP Antibody (19F461), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-56567AF750
Product Code. 18002484
  • € 558.00 / 0.10mL
Description
APIP Monoclonal specifically detects APIP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
APIP
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
APIP
The antibody was developed against a full-length His-tagged recombinant APIP.
0.1 mL
Primary
This monoclonal antibody can be used for detection of APIP and APIP2.
Store at 4°C in the dark.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
19F461
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
APAF1 interacting protein, APAF1-interacting protein, APIP2, CGI29, CGI-29, dJ179L10.2, EC 4.2.1.109, MMRP19, MTRu-1-P dehydratase, probable methylthioribulose-1-phosphate dehydratase
Mouse
Protein G purified
Apoptosis
51074.0
Human
IgG2b κ