missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APJ/Apelin receptor Antibody (72133R), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8561PEATT594
This item is not returnable.
View return policy
Description
APJ/Apelin receptor Monoclonal antibody specifically detects APJ/Apelin receptor in Human samples. It is validated for Flow Cytometry
Specifications
| APJ/Apelin receptor | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 72133R | |
| Flow Cytometry | |
| AGTRL1HG11, angiotensin II receptor-like 1, Angiotensin receptor-like 1, apelin receptor, APJ (apelin) receptor, APJ receptor, APJFLJ96609, APJR, FLJ90771, G protein-coupled receptor APJ, G-protein coupled receptor APJ, G-protein coupled receptor HG11, HG11 orphan receptor, MGC45246 | |
| Mouse myeloma cell line NS0-derived recombinant human APJ, Met1-Asp380, Accession # P35414 | |
| 0.1 mL | |
| GPCR | |
| 187 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction