Learn More
Invitrogen™ APLF Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5114861
Product Code. 17267623
  • € 497.00 / 100µL
Description
Antibody detects endogenous levels of total APLF.
C2ORF13 is a component of the cellular response to chromosomal DNA single- and double-strand breaks.[supplied by OMIM, Mar 2008].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
APLF
Polyclonal
Unconjugated
APLF
2010301N04Rik; AI452191; APFL; APLF; aprataxin- and PNK-like factor; Aprataxin and PNK-like factor; aprataxin and PNKP like factor; apurinic-apyrimidinic endonuclease APLF; C2orf13; PALF; PNK and APTX-like FHA domain-containing protein; PNK and APTX-like FHA protein; RGD1565557; XIP1; XRCC1-interacting protein 1; ZCCHH1; zinc finger, CX5CX6HX5H motif containing 1
Rabbit
Affinity chromatography
RUO
200558, 500247, 72103
-20°C
Liquid
Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide
Q8IW19, Q9D842
APLF
A synthesized peptide derived from human APLF(Accession Q8IW19), corresponding to amino acid residues L92-G142.
100 μL
Primary
Human, Mouse, Rat