missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ APLP2 Recombinant Rabbit Monoclonal Antibody (23GB1595), Invitrogen™
GREENER_CHOICE

Product Code. p-200087755
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30088661 100 μL 100µL
30088007 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30088661 Supplier Invitrogen™ Supplier No. MA553664

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

APLP2 May play a role in the regulation of hemostasis. The soluble form may have inhibitory properties towards coagulation factors. May interact with cellular G-protein signaling pathways. May bind to the DNA 5'-GTCACATG-3' (CDEI box).
TRUSTED_SUSTAINABILITY

Specifications

Antigen APLP2
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1595
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene APLP2
Gene Accession No. Q06481
Gene Alias AI790698; amyloid beta (A4) precursor-like protein 2; amyloid beta (A4) precursor-like protein 2-like; amyloid beta precursor like protein 2; amyloid precursor protein homolog HSD-2; Amyloid protein homolog; Amyloid protein precursor-like protein 2; amyloid-like protein 2; APLP2; APLP-2; APPH; APPL2; CDEBP; CDEI box-binding protein; sperm membrane protein (YWK-II); sperm membrane protein YWK-II; testicular tissue protein Li 23; WALPLP2
Gene Symbols APLP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human APLP2 (221-280AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 334
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.