missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APMAP Antibody (OTI4F6), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72080MFV450
This item is not returnable.
View return policy
Description
APMAP Monoclonal antibody specifically detects APMAP in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry
Specifications
| APMAP | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Purified |
| Flow Cytometry | |
| OTI4F6 | |
| Flow Cytometry | |
| adipocyte plasma membrane-associated protein, APMAP, BSCv, chromosome 20 open reading frame 3, Protein BSCv | |
| Full length human recombinant protein of human C20orf3(NP_065392) produced in HEK293T cell. | |
| 0.1 mL | |
| Signal Transduction | |
| 57136 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction