Learn More
PeproTech ApoA1 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Descripción
AA Sequence of recombinant protein: MDEPPQSPWD RVKDLATVYV DVLKDSGRDY VSQFEGSALG KQLNLKLLDN WDSVTSTFSK LREQLGPVTQ EFWDNLEKET EGLRQEMSKD LEEVKAKVQP YLDDFQKKWQ EEMELYRQKV EPLRAELQEG ARQKLHELQE KLSPLGEEMR DRARAHVDAL RTHLAPYSDE LRQRLAARLE ALKENGGARL AEYHAKATEH LSTLSEKAKP ALEDLRQGLL PVLESFKVSF LSALEEYTKK LNTQ. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human ApoA-I. Anti-Human ApoA-I-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human ApoA-I by sandwich ELISA (using 100 μL/well), a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human ApoA-I (500-P331) as a capture antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human ApoA-I. Western Blot: To detect Human ApoA-I by Western Blot analysis, this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible development reagents, the detection limit for Recombinant Human ApoA-I is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P331BT-1MG will be provided as 2 x 500 μg
Especificaciones
Especificaciones
| Antigen | ApoA1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1-1.0 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with no preservative |
| Gene | APOA1 |
| Gene Accession No. | P02647 |
| Gene Alias | 1-Sep; 2-Sep; Alp-1; Apo a; APO A1; apo A-1; APO I; apo(a); APOA1; Apoa-1; ApoAI; Apo-AI; apoA-I; Apolipoprotein; Apolipoprotein A1; apolipoprotein A-1; Apolipoprotein AI; apolipoprotein A-I; apolipoprotein A-I precursor; Apolipoprotein A-I(1-242); Brp-14; Ltw-1; Lvtw-1; MGC102525; MGC117399; MGC124929; preproapolipoprotein A-I; prepro-apolipoprotein-AI; ProapoA-I; Proapolipoprotein A-I; Sep-1; Sep2; Sep-2; Truncated apolipoprotein A-I; Udpgt; Ugt1; Ugt1a7 |
| Gene Symbols | APOA1 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.