missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ApoA2 Monoclonal Antibody (395906)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA561404

Product Code. 30536065

  • € 471.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

At least 9 distinct polymorphic forms of apolipoproteins are known. The apolipoproteins act as stabilizers of the intact lipoprotein particles. Quantitative measurements of HDL, LDL, and VLDL particles of human serum are often used to estimate an individual's relative risk of coronary heart disease. In addition, quantitative immunological measurements of certain apolipoproteins ( especially A-1 and B) have been suggested to be more accurate estimators of coronary heart disease that measurements of lipoprotein particles (especially HDL and LDL). The apolipoproteins are derived from human plasma by density gradient ultracentrifugation and HPLC.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ApoA2
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P02652
APOA2
E. coli-derived recombinant human ApoA2, Gln24-Gln100.
100 μg
Primary
Human
Antibody
IgG2a
Western Blot
395906
Unconjugated
APOA2
Alp-2; APO A2; APOA2; Apoa-2; ApoAII; Apo-AII; ApoA-II; Apolipoprotein; Apolipoprotein A2; Apolipoprotein AII; apolipoprotein A-II; Apolipoprotein A-II(1-76); Hdl-1; ProapoA-II; Proapolipoprotein A-II; Truncated apolipoprotein A-II
Rat
Protein A/G
RUO
336
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.