missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOA5 Monoclonal Antibody (OTI2B11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA507101
This item is not returnable.
View return policy
Description
Apolipoprotein A4 (also known as ApoA-IV) is a plasma protein that is O-linked glycoprotein after proteolytic processing. ApoA4 is a major constituent of HDL and it also can be found on the surface of newly synthesized chylomicrons. The physiological function of ApoA4 is not completely identified so far, but several evidences from vitro studies indicate that this apolipoprotein involves in several steps of the reverse cholesterol transport pathway, which transports cholesterol from peripheral cells back to the liver.Specifications
| APOA5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6Q788 | |
| APOA5 | |
| Full length human recombit protein of human APOA5 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2B11 | |
| Unconjugated | |
| APOA5 | |
| APOAV, RAP3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 116519 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction