missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOB48R Antibody (OTI1B1), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72052IR
This item is not returnable.
View return policy
Description
APOB48R Monoclonal specifically detects APOB48R in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| APOB48R | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| APOBR | |
| Recombinant protein fragment corresponding to amino acids 780-1088 of human APOB48R (NP_061160) produced in E.coli. | |
| 0.1 mL | |
| 55911.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1B1 | |
| Western Blot, Immunohistochemistry | |
| APOB100R, apoB-48R, APOB48Rapolipoprotein B48 receptor, apolipoprotein B receptor, Apolipoprotein B-100 receptor, Apolipoprotein B-48 receptor | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction