Learn More
LSP1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP3-00254PCP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
LSP1 Polyclonal antibody specifically detects LSP1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Spécification
| Western Blot, Immunohistochemistry (Paraffin) | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Store at 4C in the dark. | |
| Purified |