missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ APOBEC3G Polyclonal Antibody
GREENER_CHOICE

Product Code. 12888262
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12888262 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12888262 Supplier Invitrogen™ Supplier No. PA520052

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is Caco-2 cell lysate. PA5-20052 can be used with blocking peptide PEP-0171.

The Apoliprotein B mRNA-editing, enzyme-catalytic, polypeptide-like (APOBEC) 3 is a multi-isoform member of the cytidine deaminase family of enzymes that act on monomeric nucleoside and nucleotide substrates. Similar to TRIM5-a which targets incoming retroviral capsids, APOBEC3 plays a major role in cellular defense against retroviral infection as at least two isoforms, APOBEC3G and to a lesser extent APOBEC3F, can be incorporated HIV-1 virions and induce hypermutation in the newly synthesized viral DNA and thus destabilize the viral genome. This innate mechanism of retroviral resistance is counteracted by the HIV-1 Vif protein by inducing the ubiquitization and degradation of APOBEC3G; a single amino acid substitution (D128K) blocks APOBEC3G depletion without affecting its inhibitory activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APOBEC3G
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene APOBEC3G
Gene Accession No. Q9HC16
Gene Alias A3G; APOBEC3G; APOBEC-related cytidine deaminase; APOBEC-related protein; APOBEC-related protein 9; apolipoprotein B editing enzyme catalytic polypeptide-like 3G; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3G; apolipoprotein B mRNA editing enzyme catalytic subunit 3G; apolipoprotein B mRNA editing enzyme cytidine deaminase; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3G; apolipoprotein B mRNA-editing enzyme catalytic polypeptide 3G; ARCD; ARP9; ARP-9; ASGPR2; ASGP-R2; bK150C2.7; CEM15; CEM-15; CLEC4H2; Deoxycytidine deaminase; dJ494G10.1; DNA dC->dU editing enzyme; DNA dC->dU-editing enzyme APOBEC-3G; FLJ12740; HBXBP; HL-2; MDS019; OTTHUMP00000028911; phorbolin-like protein MDS019
Gene Symbols APOBEC3G
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 15 amino acids near the amino-terminus of human APOBEC3G.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60489
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.