Learn More
APOBEC4 Mouse anti-Human, Clone: OTI3F11, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF810779
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the AID/APOBEC family of polynucleotide (deoxy)cytidine deaminases, which convert cytidine to uridine. Other AID/APOBEC family members are involved in mRNA editing, somatic hypermutation and recombination of immunoglobulin genes, and innate immunity to retroviral infection.Specifications
| APOBEC4 | |
| Monoclonal | |
| Unconjugated | |
| APOBEC4 | |
| C1orf169 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 403314 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI3F11 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q8WW27 | |
| APOBEC4 | |
| Full length human recombit protein of human APOBEC4 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.