Learn More
APOH Mouse anti-Human, Clone: OTI10A6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF807230
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Apolipoprotein H has been implicated in a variety of physiologic pathways including lipoprotein metabolism, coagulation, and the production of antiphospholipid autoantibodies. APOH may be a required cofactor for anionic phospholipid binding by the antiphospholipid autoantibodies found in sera of many patients with lupus and primary antiphospholipid syndrome, but it does not seem to be required for the reactivity of antiphospholipid autoantibodies associated with infections.Specifications
| APOH | |
| Monoclonal | |
| Unconjugated | |
| APOH | |
| B2G1, B2GP1, BG | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 350 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI10A6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P02749 | |
| APOH | |
| Human recombit protein fragment corresponding to amino acids 120-345 of human APOH produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.