missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOL1 Monoclonal Antibody (OTI5E10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812043
This item is not returnable.
View return policy
Description
APOL1 is a secreted high density lipoprotein which binds to apolipoprotein A-I. Apolipoprotein A-I is a relatively abundant plasma protein and is the major apoprotein of HDL. It is involved in the formation of most cholesteryl esters in plasma and also promotes efflux of cholesterol from cells. This apolipoprotein L family member may play a role in lipid exchange and transport throughout the body, as well as in reverse cholesterol transport from peripheral cells to the liver.Specifications
| APOL1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O14791 | |
| APOL1 | |
| Human recombit protein fragment corresponding to amino acids 28-339 of human APOL1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5E10 | |
| Unconjugated | |
| APOL1 | |
| APO-L, APOL, APOL-I, FSGS4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8542 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction