Learn More
APOL1-Specific Mouse anti-Human, CoraLite 594, Clone: 1G12D11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66124-100UL
Description
The APOL1-Specific antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human APOL1-Specific. This antibody recognizes human antigen. The APOL1-Specific antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| APOL1-Specific | |
| Monoclonal | |
| 2000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Immunofluorescence | |
| 1G12D11 | |
| CoraLite 594 | |
| 100 μL | |
| 8542 | |
| IgG2a |