Learn More
APOL1-Specific Mouse anti-Human, CoraLite 594, Clone: 1G12D11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL594-66124-100UL
Product Code. 16880966
  • € 450.00 / 100µL
Description
The APOL1-Specific antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human APOL1-Specific. This antibody recognizes human antigen. The APOL1-Specific antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
Specifications
APOL1-Specific
Monoclonal
2000 μg/mL
Mouse
Primary
Human
Immunofluorescence
1G12D11
CoraLite 594
100 μL
8542