missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein A-I/ApoA1 Antibody (2083D), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals MAB3664B
This item is not returnable.
View return policy
Description
Apolipoprotein A-I/ApoA1 Monoclonal specifically detects Apolipoprotein A-I/ApoA1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| Apolipoprotein A-I/ApoA1 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| APOA1 | |
| rhApoA1 aa25-267 | |
| 100 μL | |
| Breast Cancer, Cancer, Diabetes Research, Lipid and Metabolism | |
| 335 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| 2083D | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| APOA1, apoA-I, Apo-AI, Apolipoprotein A1, apolipoprotein A-I, MGC117399 | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction