missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein A-I/ApoA1 Rabbit anti-Human, Clone: 3, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 379.00 - € 552.00
Specifications
| Antigen | Apolipoprotein A-I/ApoA1 |
|---|---|
| Clone | 3 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18247276
|
Novus Biologicals
NBP2-89598-50UL |
50 μg |
€ 379.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18207677
|
Novus Biologicals
NBP2-89598-100UL |
100 μg |
€ 552.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Apolipoprotein A-I/ApoA1 Monoclonal specifically detects Apolipoprotein A-I/ApoA1 in Human samples. It is validated for ELISA.Specifications
| Apolipoprotein A-I/ApoA1 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| APOA1, Apo-AI, ApoA-I, Apolipoprotein A1, apolipoprotein A-I, MGC117399 | |
| Recombinant Human Apolipoprotein A-I/ApoA1 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 3 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Biologically Active Proteins, Breast Cancer, Cancer, Diabetes Research, Lipid and Metabolism | |
| 0.2 um filtered solution in PBS | |
| 335 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title