missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Apolipoprotein A4 Recombinant Rabbit Monoclonal Antibody (3M5S4)

Product Code. 30130951
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30130951 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30130951 Supplier Invitrogen™ Supplier No. MA555246

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen Sequence: EVSADQVATV MWDYFSQLSN NAKEAVEHLQ KSELTQQLNA LFQDKLGEVN TYAGDLQKKL VPFATELHER LAKDSEKLKE EIGKELEELR ARLLPHANEV SQKIGDNLRE LQQRLEPYAD QLRTQVSTQA EQLRRQLTPY AQRMERVLRE NADSLQASLR PHADELKAKI DQNVEELKGR LTPYADEFKV KIDQTVEELR RSLAPYAQDT QEKLNHQLEG LTFQMKKNAE ELKARISASA EELRQRLAPL AEDVRGNLRG NTEGLQKSLA ELGGHLDQQV EEFRRRVEPY GENFNKALVQ QMEQLRQKLG PHAGDVEGHL SFLEKDLRDK VNSFFSTFKE KESQDKTLSL PELEQQQEQQ QEQQQEQVQM LAPLES.

Apolipoprotein A4 (also known as ApoA-IV) is a plasma protein that is an O-linked glycoprotein after proteolytic processing. ApoA4 is a major constituent of HDL and it also can be found on the surface of newly synthesized chylomicrons. The physiological function of ApoA4 is not completely identified so far, but evidence from several in vitro studies indicate that this apolipoprotein is involved in several steps of the reverse cholesterol transport pathway, which transports cholesterol from peripheral cells back to the liver.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein A4
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 3M5S4
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene APOA4
Gene Accession No. P06727
Gene Alias APOA4; Apoa-4; APOAIV; apo-AIV; ApoA-IV; Apolipoprotein A4; apolipoprotein AIV; apolipoprotein A-IV; MGC142154; MGC142156
Gene Symbols APOA4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 21-396 of human APOA4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 337
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.