missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein B/ApoB Antibody (APOB/3300), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08226G
This item is not returnable.
View return policy
Description
Apolipoprotein B/ApoB Monoclonal antibody specifically detects Apolipoprotein B/ApoB in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications
| Apolipoprotein B/ApoB | |
| Monoclonal | |
| DyLight 488 | |
| Apo B-100, apoB-100, apoB-48, apolipoprotein B (including Ag(x) antigen), apolipoprotein B-100, apolipoprotein B48, FLDB, LDLCQ4, mutant Apo B 100 | |
| Human recombinant Apolipoprotein B/ApoB protein fragment (around aa592-689) (exact sequence is proprietary) (Uniprot: P04114) | |
| 100 μg | |
| Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
| 338 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Protein Array | |
| APOB/3300 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction