Learn More
Invitrogen™ Apolipoprotein B Monoclonal Antibody (F2C4)

Description
MA5-14740 targets Apolipoprotein B in ELISA, IP, and RIA applications and shows reactivity with Human samples. The MA5-14740 immunogen is purified human serum LDL. MA5-14740 detects Apolipoprotein B which has a predicted molecular weight of approximately 513 kDa. Product MA514740 is a smaller package size of MIA1606 (formerly sold as a Seradyn product).
Specifications
Specifications
| Antigen | Apolipoprotein B |
| Applications | ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Radioimmune Assays (RIA), Immunocytochemistry |
| Classification | Monoclonal |
| Clone | F2C4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 |
| Gene | APOB |
| Gene Accession No. | P04114 |
| Gene Alias | Aa1064; Ac1-060; AI315052; Apo B100; Apo B-100; Apo B-48; APOB; apo-B; ApoB 100; ApoB 48; ApoB-100; apob-48; apolipo b; apolipoprotein B; apolipoprotein B (including Ag(x) antigen); Apolipoprotein B 100; Apolipoprotein B 48; apolipoprotein B PI; Apolipoprotein B100; apolipoprotein B-100; apolipoprotein B46; apolipoprotein B47; apolipoprotein B48; Apolipoprotein B-48; apolipoprotein B49; FLDB; LDLCQ4; LOX-1; mCG_129875; MGC176318; Ox-LDL receptor 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.