missing translation for 'onlineSavingsMsg'
Learn More

Apolipoprotein C-II/ApoC2 Antibody, Novus Biologicals™

Product Code. 18199799 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.2 mg
Unit Size:
0.2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18199799 0.2 mg 0.2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18199799 Supplier Novus Biologicals Supplier No. NB400155

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Apolipoprotein C-II/ApoC2 Polyclonal specifically detects Apolipoprotein C-II/ApoC2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein C-II/ApoC2
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:10000, ELISA 1:5000-1:10000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:50-1:200
Formulation 0.125 M Sodium Borate, 0.075 M Sodium Chloride, 0.005 M EDTA, pH 8.0 with 0.01% Sodium Azide
Gene Accession No. P02655
Gene Alias APC2, APOC2, Apo-CII, ApoC-II, Apolipoprotein C2, apolipoprotein C-II, MGC75082
Gene Symbols APOC2
Host Species Goat
Immunogen Apolipoprotein C-II/ApoC2 produced synthetically in full-length form (not selected epitopes) using conventional peptide technology. (Uniprot: P02655)
Molecular Weight of Antigen 11.28 kDa
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 344
Test Specificity Typically less than 1% cross reactivity against other types of apoLipoprotein was detected by ELISA against purified standards. This antibody reacts with human apoLipoprotein C-II and has negligible cross-reactivity with Type A-I, A-II, B, C-I, C-III, E and J apoLipoproteins. Specific cross reaction of anti-apoLipoprotein antibodies with antigens from other species has not been determined. Non-specific cross reaction of anti-apoLipoprotein antibodies with other human serum proteins is negligible.
Target Species Human
Content And Storage Store at 4°C short term. For extended storage, add an equal volume of glycerol, aliquot and store at -20°C or below. Avoid repeated freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.