missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein CIII Antibody (SR1078), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | Apolipoprotein CIII |
|---|---|
| Clone | SR1078 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18682845
|
Novus Biologicals
NBP3-21769-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18689923
|
Novus Biologicals
NBP3-21769-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Apolipoprotein CIII Monoclonal antibody specifically detects Apolipoprotein CIII in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| Apolipoprotein CIII | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| APOC3, apo-CIII, ApoC-III, APOCIII, Apolipoprotein C3, apolipoprotein C-III, MGC150353 | |
| A synthesized peptide derived from human Apolipoprotein CIII (Uniprot #: P02656) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1078 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 345 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title