Learn More
Apolipoprotein D Antibody (APOD/3412), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-08417APC
Product Code. 18661369
  • € 690.00 / 0.10mL
Description
Apolipoprotein D Monoclonal antibody specifically detects Apolipoprotein D in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Apolipoprotein D
Monoclonal
APC
apo-D, ApoD, apolipoprotein D
Human recombinant Apolipoprotein D protein fragment (around aa 21-119) (exact sequence is proprietary) (Uniprot: P05090)
0.1 mL
Alzheimers Research, Cancer, Cardiovascular Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
347
Store at 4°C in the dark.
IgG2b κ
Immunohistochemistry (Paraffin), Protein Array
APOD/3412
PBS
Mouse
Protein A or G purified