missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein D Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-40025
This item is not returnable.
View return policy
Description
Apolipoprotein D Polyclonal antibody specifically detects Apolipoprotein D in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Apolipoprotein D | |
| Polyclonal | |
| Unconjugated | |
| PBS and 50% Glycerol | |
| apo-D, ApoD, apolipoprotein D | |
| Synthetic peptide from mouse Apolipoprotein D. | |
| 0.05 mg | |
| Alzheimers Research, Cancer, Cardiovascular Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 347 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 1 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 10 μg/mL | |
| P51910 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction