missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein E R2/ApoE R2 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2217DL594
This item is not returnable.
View return policy
Description
Apolipoprotein E R2/ApoE R2 Polyclonal antibody specifically detects Apolipoprotein E R2/ApoE R2 in Human, Mouse, Bovine, Chicken samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Apolipoprotein E R2/ApoE R2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Bovine, Chicken | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| DyLight 594 | |
| APOER2HSZ75190, Apolipoprotein E receptor 2, low density lipoprotein receptor-related protein 8, apolipoprotein e receptor, low-density lipoprotein receptor-related protein 8, LRP-8, MCI1 | |
| A synthetic peptide made to a C-terminal portion of the human ApoER2 protein sequence (between residues 800-900). [UniProt# Q14114] | |
| 0.1 mL | |
| Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 7804 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction