missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein H/ApoH Antibody (APOH/3705) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-26956JF669
This item is not returnable.
View return policy
Description
Apolipoprotein H/ApoH Monoclonal antibody specifically detects Apolipoprotein H/ApoH in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Apolipoprotein H/ApoH | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Activated protein C-binding protein, Anticardiolipin cofactor, APC inhibitor, Apo-H, Apolipoprotein H, apolipoprotein H (beta-2-glycoprotein I), B2G1, B2GP1, B2GPI, Beta(2)GPI, beta-2-glycoprotein 1, Beta-2-glycoprotein I, BG | |
| Human recombinant Apolipoprotein H/ApoH protein fragment (around aa 592-689) (exact sequence is proprietary) | |
| 0.1 mL | |
| Cholesterol Metabolism, Lipid and Metabolism | |
| 350 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| APOH/3705 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction