missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Apolipoprotein H/ApoH Antibody (APOH/3705), Novus Biologicals™

Mouse Monoclonal Antibody

€ 245.00 - € 486.00

Specifications

Antigen Apolipoprotein H/ApoH
Clone APOH/3705
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043867
View Documents
Novus Biologicals™
NBP3-26808-20ug
20 μg
€ 245.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043379
View Documents
Novus Biologicals™
NBP3-26808-100ug
100 μg
€ 486.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Apolipoprotein H/ApoH Monoclonal antibody specifically detects Apolipoprotein H/ApoH in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Apolipoprotein H/ApoH
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cholesterol Metabolism, Lipid and Metabolism
10 mM PBS with 0.05% BSA
350
IgG1 κ
Protein A or G purified
APOH/3705
Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
Monoclonal
Purified
RUO
Human
Activated protein C-binding protein, Anticardiolipin cofactor, APC inhibitor, Apo-H, Apolipoprotein H, apolipoprotein H (beta-2-glycoprotein I), B2G1, B2GP1, B2GPI, Beta(2)GPI, beta-2-glycoprotein 1, Beta-2-glycoprotein I, BG
Human recombinant Apolipoprotein H/ApoH protein fragment (around aa 592-689) (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.