missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein L1 Antibody (CL0170), Novus Biologicals™
Mouse Monoclonal Antibody
€ 369.00 - € 529.00
Specifications
| Antigen | Apolipoprotein L1 |
|---|---|
| Clone | CL0170 |
| Dilution | Western Blot 1 μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18613889
|
Novus Biologicals
NBP2-52870-25ul |
25 μL |
€ 369.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18637088
|
Novus Biologicals
NBP2-52870 |
0.1 mL |
€ 529.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Apolipoprotein L1 Monoclonal antibody specifically detects Apolipoprotein L1 in Human samples. It is validated for Western BlotSpecifications
| Apolipoprotein L1 | |
| Western Blot 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ApoL, APO-L, ApoL-I, Apolipoprotein L, apolipoprotein L, 1, apolipoprotein L1, Apolipoprotein L-I, FSGS4 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: SNFLSLAGNTYQLTRGIGKDIRALRRARANLQSVPHASASRPRVTEPISAESGEQVERVNEPSILEMSRGVKLTDVAPVSFFLVLDVVYLVYESKHLHEGAKSETAEELKKVAQELEEKLNILNN | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL0170 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cancer, Cardiovascular Biology, Endocrinology | |
| PBS (pH 7.2), 40% Glycerol | |
| 8542 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title