Learn More
Invitrogen™ Apolipoprotein L2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5101432
Product Code. 16697904
  • € 493.00 / 100µL
Description
Antibody detects endogenous levels of total APOL2.
This gene is a member of the apolipoprotein L gene family. The encoded protein is found in the cytoplasm, where it may affect the movement of lipids or allow the binding of lipids to organelles. Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Apolipoprotein L2
Polyclonal
Unconjugated
APOL2
Apol2; ApoL-2; APOL3; Apol8; APOL-II; apolipoprotein L 8; apolipoprotein L, 2; Apolipoprotein L2; Apolipoprotein L-2; apolipoprotein L-II; OTTHUMP00000028936
Rabbit
Affinity Chromatography
RUO
23780, 315111
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9BQE5
APOL2
A synthesized peptide derived from human APOL2(Accession Q9BQE5), corresponding to amino acid residues E305-Q337.
100 μL
Primary
Human, Rat