missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOOL Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99175B
This item is not returnable.
View return policy
Description
APOOL Polyclonal antibody specifically detects APOOL in Human samples. It is validated for Western Blot
Specifications
| APOOL | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 139322 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| AAIR8193, apolipoprotein O-like, chromosome X open reading frame 33, CXorf33, FAM121A, family with sequence similarity 121A, MGC129748, Protein FAM121A, UNQ8193 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human APOOL. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction