missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thyroglobulin Antibody (TGB/1968R), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08269F
This item is not returnable.
View return policy
Description
Thyroglobulin Monoclonal antibody specifically detects Thyroglobulin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Thyroglobulin | |
| Monoclonal | |
| FITC | |
| AITD3TGN, TDH3, Tg, thyroglobulin | |
| Recombinant full-length human Thyroglobulin protein (Uniprot: P01266) | |
| 100 μg | |
| Cancer, Cell Biology | |
| 7038 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| TGB/1968R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction