Learn More
APP Antibody (6E10) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody has been used in 2 publications
Brand:  Novus Biologicals NBP2-62566-0.025MG
Product Code. 18076304
  • € 304.00 / 25µg
Description
APP Monoclonal specifically detects APP in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Electron Microscopy.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
APP
Monoclonal
Unconjugated
PBS
APP
Beta Amyloid signal peptide 1-16.
351
Human
Purified
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunomicroscopy
60000000000
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin, Electron Microscopy 1:10 - 1:500
amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP
Rabbit
0.025 mg
This antibody binds to amino acid residues 1-16 of beta amyloid.
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
IgG κ